Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GGA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001560.2 | NP_001001560.1 | 2 | 471 | ||
NM_001172687.1 | NP_001166158.1 | 2 | 471 | |||
NM_001172688.1 | NP_001166159.1 | 3 | 517 | |||
NM_013365.4 | NP_037497.1 | 2 | 471 | |||
XM_005261517.3 | XP_005261574.1 | 3 | 645 | |||
XM_005261518.1 | XP_005261575.1 | 2 | 156 | |||
XM_005261520.1 | XP_005261577.1 | 2 | 362 | |||
XM_006724229.1 | XP_006724292.1 | 3 | 404 | |||
XM_011530122.1 | XP_011528424.1 | 3 | 578 | |||
XM_011530123.1 | XP_011528425.1 | 2 | 156 | |||
XM_011530124.1 | XP_011528426.1 | 3 | 645 | |||
XM_017028760.1 | XP_016884249.1 | 3 | 645 | |||
XM_017028761.1 | XP_016884250.1 | 2 | 362 | |||
XM_017028762.1 | XP_016884251.1 | 3 | 645 | |||
XM_017028763.1 | XP_016884252.1 | 2 | 111 | |||
XM_017028764.1 | XP_016884253.1 | 2 | 362 | |||
GenBank mRNA | AF190862.1 | - | 2 | 99 | ||
AF218584.1 | - | 2 | 88 | |||
AF233521.1 | - | 2 | 106 | |||
AK075256.1 | - | 2 | 449 | |||
AK122898.1 | - | 3 | 517 | |||
AK225157.1 | - | 2 | 105 | |||
AK290548.1 | - | 2 | 110 | |||
BC010917.1 | - | 2 | 115 | |||
BC029388.1 | - | 2 | 110 | |||
BC044629.1 | - | 2 | 100 | 5796877 | ||
CR456493.1 | - | 2 | 220 |