Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015603.2 | NP_056418.1 | 8, 9 | 1100 | ||
XM_017026578.1 | XP_016882067.1 | 8, 9 | 1145 | |||
GenBank mRNA | AK302153.1 | - | 9, 10 | 1021 | ||
AL050284.1 | - | 8, 9 | 1039 | |||
BC002787.2 | - | 8, 9 | 996 | 3615928 | ||
BC009743.2 | - | 8, 9 | 996 | 3832697 |