Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TANC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025185.3 | NP_079461.2 | 24 | 3984 | ||
XM_005257203.4 | XP_005257260.1 | 25 | 4211 | |||
XM_006721806.3 | XP_006721869.1 | 27 | 4463 | |||
XM_006721807.3 | XP_006721870.1 | 26 | 4433 | |||
XM_011524597.2 | XP_011522899.1 | 27 | 4463 | |||
XM_011524598.2 | XP_011522900.1 | 26 | 4433 | |||
XM_011524599.1 | XP_011522901.1 | 27 | 4289 | |||
XM_011524600.2 | XP_011522902.1 | 26 | 4352 | |||
XM_011524601.2 | XP_011522903.1 | 27 | 4352 | |||
XM_011524602.2 | XP_011522904.1 | 26 | 4241 | |||
XM_017024429.1 | XP_016879918.1 | 26 | 4259 | |||
XM_017024430.1 | XP_016879919.1 | 23 | 7630 | |||
GenBank mRNA | AB046856.2 | - | 20 | 3585 | ||
AJ278120.1 | - | 11 | 1388 |