Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIF26A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015656.1 | NP_056471.1 | 12 | 4032 | ||
XM_011536641.2 | XP_011534943.1 | 9 | 3177 | |||
GenBank mRNA | AB033062.2 | - | 11 | 3906 | ||
AB290161.1 | - | 11 | 4002 | |||
BC025353.1 | - | 1 | 183 |