Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIMM13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012458.3 | NP_036590.1 | 2, 3 | 560 | ||
GenBank mRNA | AF144700.1 | - | 2, 3 | 223 | ||
AF152352.1 | - | 2, 3 | 177 | |||
AK024764.1 | - | 2, 3 | 195 | |||
BC008607.1 | - | 2, 3 | 218 | 3453921 | ||
BI908100.1 | - | 2, 3 | 521 | |||
DA890574.1 | - | 2, 3 | 532 | |||
DA947460.1 | - | 567 |