Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIMM13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012458.3 | NP_036590.1 | 3 | 623 | ||
GenBank mRNA | AF144700.1 | - | 3 | 286 | ||
AF152352.1 | - | 3 | 240 | |||
AK024764.1 | - | 3 | 258 | |||
BC008607.1 | - | 3 | 281 | 3453921 | ||
BI908100.1 | - | 3 | 584 |