Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STAU2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164380.1 | NP_001157852.1 | 11 | 1467 | ||
NM_001164381.1 | NP_001157853.1 | 10 | 1336 | |||
NM_001164382.1 | NP_001157854.1 | 9 | 1176 | |||
NM_001164383.1 | NP_001157855.1 | 6 | 861 | |||
NM_001164384.1 | NP_001157856.1 | 9 | 1400 | |||
NM_001164385.1 | NP_001157857.1 | 8 | 1157 | |||
NM_014393.2 | NP_055208.2 | 10 | 1336 | |||
GenBank mRNA | AB210014.1 | - | 10 | 1232 | ||
AF459097.1 | - | 9 | 1186 | |||
AF459098.1 | - | 8 | 1055 | |||
AK001576.1 | - | 3 | 362 | |||
AK002152.1 | - | 11 | 1380 | |||
AK023314.1 | - | 2 | 239 | |||
AK290141.1 | - | 9 | 1400 | |||
AK293496.1 | - | 6 | 774 | |||
AK293729.1 | - | 8 | 1097 | |||
AK294466.1 | - | 8 | 1109 | |||
AK303104.1 | - | 9 | 1089 | |||
AK307841.1 | - | 6 | 797 | |||
AK314617.1 | - | 10 | 1249 | |||
BC008369.1 | - | 10 | 1249 | 3682739 | ||
BC110447.1 | - | 8 | 1073 | 40009098 | ||
CR627442.1 | - | 6 | 843 | |||
Y19062.1 | - | 10 | 1231 |