Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STAU2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164380.1 | NP_001157852.1 | 7 | 879 | ||
NM_001164381.1 | NP_001157853.1 | 6 | 748 | |||
NM_001164382.1 | NP_001157854.1 | 5 | 588 | |||
NM_001164383.1 | NP_001157855.1 | 2 | 273 | |||
NM_001164384.1 | NP_001157856.1 | 5 | 812 | |||
NM_001164385.1 | NP_001157857.1 | 4 | 569 | |||
NM_014393.2 | NP_055208.2 | 6 | 748 | |||
GenBank mRNA | AB210014.1 | - | 6 | 644 | ||
AF459097.1 | - | 5 | 598 | |||
AF459098.1 | - | 4 | 467 | |||
AK002152.1 | - | 7 | 792 | |||
AK290141.1 | - | 5 | 812 | |||
AK293496.1 | - | 2 | 186 | |||
AK293729.1 | - | 4 | 509 | |||
AK294466.1 | - | 4 | 521 | |||
AK303104.1 | - | 5 | 501 | |||
AK307841.1 | - | 2 | 208 | |||
AK314617.1 | - | 6 | 661 | |||
BC008369.1 | - | 6 | 661 | 3682739 | ||
BC110447.1 | - | 4 | 485 | 40009098 | ||
CR627442.1 | - | 2 | 230 | |||
Y19062.1 | - | 6 | 643 |