Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARRDC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025604.2 | NP_001020775.1 | 7, 8 | 1335 | ||
NM_015683.1 | NP_056498.1 | 7, 8 | 1310 | |||
GenBank mRNA | AF131826.1 | - | 6, 7 | 842 | ||
AF193051.1 | - | 7, 8 | 1310 | |||
AK127940.1 | - | 7, 8 | 1299 | |||
AK128588.1 | - | 1, 2 | 2557 | |||
AK314657.1 | - | 7, 8 | 1308 | |||
BC022516.1 | - | 7, 8 | 1308 | 4817429 |