Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDCBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199784.1 | NP_001186713.1 | 6 | 557 | ||
NM_015685.5 | NP_056500.2 | 2 | 448 | |||
NM_080489.4 | NP_536737.3 | 6 | 668 | |||
GenBank mRNA | AF131809.1 | - | 2 | 414 | ||
AF159228.1 | - | 6 | 557 | |||
AJ292244.1 | - | 5 | 499 | |||
AJ292245.1 | - | 2 | 336 | |||
AK022318.1 | - | 1 | 2623 | |||
BC002727.2 | - | 6 | 607 | 3633262 | ||
BI599848.1 | - | 2 | 452 | |||
CB154525.1 | - | 2 | 434 |
Gene Symbol: FKBP1A-SDCBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037661.1 | 7 | 776 |