Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DKKL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001197301.1 | NP_001184230.1 | 2 | 317 | ||
NM_001197302.1 | NP_001184231.1 | 2 | 278 | |||
NM_014419.3 | NP_055234.1 | 2 | 317 | |||
XM_006723142.2 | XP_006723205.1 | 2 | 163 | |||
XM_011526724.1 | XP_011525026.1 | 2 | 187 | |||
XM_011526725.1 | XP_011525027.1 | 2 | 137 | |||
XM_011526726.2 | XP_011525028.1 | 2 | 397 | |||
XM_011526727.2 | XP_011525029.1 | 2 | 187 | |||
XM_011526728.2 | XP_011525030.1 | 2 | 187 | |||
XM_011526729.1 | XP_011525031.1 | 2 | 158 | |||
XM_017026593.1 | XP_016882082.1 | 2 | 170 | |||
GenBank mRNA | AF177398.1 | - | 2 | 168 | ||
AI879324.1 | - | 2 | 507 | |||
AY358922.1 | - | 2 | 317 | |||
BC030581.2 | - | 2 | 197 | 4836991 | ||
BI826695.1 | - | 2 | 278 | |||
BI826744.1 | - | 2 | 168 |