Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DKKL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001197301.1 | NP_001184230.1 | 4 | 751 | ||
NM_001197302.1 | NP_001184231.1 | 5 | 805 | |||
NM_014419.3 | NP_055234.1 | 5 | 844 | |||
XM_006723142.2 | XP_006723205.1 | 5 | 690 | |||
XM_011526724.1 | XP_011525026.1 | 5 | 714 | |||
XM_011526725.1 | XP_011525027.1 | 5 | 664 | |||
XM_011526726.2 | XP_011525028.1 | 5 | 924 | |||
XM_011526727.2 | XP_011525029.1 | 4 | 621 | |||
XM_011526728.2 | XP_011525030.1 | 3 | 480 | |||
XM_011526729.1 | XP_011525031.1 | 5 | 685 | |||
XM_017026593.1 | XP_016882082.1 | 5 | 697 | |||
GenBank mRNA | AF177398.1 | - | 5 | 695 | ||
AY358922.1 | - | 5 | 844 | |||
BC030581.2 | - | 5 | 724 | 4836991 |