Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DKK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001018057.1 | NP_001018067.1 | 6 | 836 | ||
NM_013253.4 | NP_037385.2 | 7 | 1004 | |||
NM_015881.5 | NP_056965.3 | 7 | 1018 | |||
XM_006718178.2 | XP_006718241.1 | 7 | 831 | |||
XM_017017553.1 | XP_016873042.1 | 7 | 965 | |||
XM_017017554.1 | XP_016873043.1 | 7 | 983 | |||
XM_017017555.1 | XP_016873044.1 | 7 | 831 | |||
GenBank mRNA | AB033421.1 | - | 6 | 797 | ||
AB034203.1 | - | 7 | 976 | |||
AB057591.1 | - | 7 | 1004 | |||
AF177396.1 | - | 6 | 816 | |||
AK090952.1 | - | 197 | ||||
AK092979.1 | - | 7 | 795 | |||
AK225156.1 | - | 6 | 788 | |||
AK289897.1 | - | 7 | 845 | |||
AK295441.1 | - | 5 | 696 | |||
AK295539.1 | - | 7 | 939 | |||
AL535720.3 | - | 6 | 835 | |||
AY358378.1 | - | 7 | 931 | |||
AY587550.1 | - | 6 | 779 | |||
BC007660.2 | - | 7 | 1017 | 3347453 | ||
BQ690088.1 | - | 1 | 83 | |||
BQ690888.1 | - | 1 | 83 |