Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INGX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002226.3 | 1 | 193 | |||
GenBank mRNA | AF149724.1 | - | 1 | 194 | ||
BC101123.1 | - | 1 | 113 | |||
BC101124.1 | - | 1 | 113 | |||
BC117297.1 | - | 1 | 18 |
Gene Symbol: TAF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_104396.1 | 1 | 258 | |||
GenBank mRNA | AJ549246.2 | - | 1 | 258 |