Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TUBG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320509.1 | NP_001307438.1 | 12 | 1553 | ||
NM_016437.2 | NP_057521.1 | 11 | 1526 | |||
XM_011524614.2 | XP_011522916.1 | 10 | 1461 | |||
XM_017024453.1 | XP_016879942.1 | 10 | 1441 | |||
XM_017024454.1 | XP_016879943.1 | 11 | 2712 | |||
XM_017024455.1 | XP_016879944.1 | 11 | 1540 | |||
XM_017024456.1 | XP_016879945.1 | 11 | 2789 | |||
XM_017024457.1 | XP_016879946.1 | 12 | 1354 | |||
GenBank mRNA | AF225971.1 | - | 11 | 1519 | ||
AK022324.1 | - | 11 | 1469 | |||
AK225134.1 | - | 11 | 1485 | |||
BC009670.2 | - | 11 | 1378 | 3891214 | ||
BC051890.1 | - | 11 | 1469 | 5726689 | ||
BC108739.1 | - | 11 | 1487 | 5242733 | ||
BE727957.1 | - | 3 | 391 |