Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL36A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014440.2 | NP_055255.1 | 4 | 860 | ||
XM_005263639.2 | XP_005263696.1 | 4 | 860 | |||
XM_011510965.1 | XP_011509267.1 | 4 | 860 | |||
XM_017003806.1 | XP_016859295.1 | 4 | 860 | |||
GenBank mRNA | AF201831.1 | - | 3 | 449 | ||
AK314149.1 | - | 4 | 863 | |||
BC107042.1 | - | 3 | 449 | 40002575 | ||
BC107043.1 | - | 3 | 449 | 40002576 |