Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014450.2 | NP_055265.1 | 4 | 425 | ||
GenBank mRNA | AJ010059.1 | - | 4 | 415 | ||
AK314758.1 | - | 4 | 406 | |||
BC102029.1 | - | 4 | 423 | 40031396 | ||
BC104491.1 | - | 4 | 423 | 40031403 | ||
BC107484.1 | - | 4 | 423 | 40031398 | ||
BM919490.1 | - | 4 | 265 | |||
JQ924063.1 | - | 3 | 499 |