Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AHDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029882.3 | NP_001025053.1 | 6 | 4955 | ||
XM_005245848.3 | XP_005245905.1 | 7 | 5190 | |||
XM_005245849.3 | XP_005245906.1 | 7 | 5295 | |||
XM_005245850.3 | XP_005245907.1 | 8 | 5315 | |||
XM_005245851.3 | XP_005245908.1 | 7 | 5187 | |||
XM_005245852.3 | XP_005245909.1 | 6 | 4964 | |||
XM_011541255.1 | XP_011539557.1 | 7 | 4869 | |||
XM_011541256.2 | XP_011539558.1 | 7 | 5309 | |||
XM_011541257.2 | XP_011539559.1 | 6 | 6064 | |||
GenBank mRNA | AK125431.1 | - | 1 | 1673 | ||
BC002677.2 | - | 1 | 751 | |||
BC014394.1 | - | 1 | 751 | |||
U79259.1 | - | 1 | 716 |