Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AHDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029882.3 | NP_001025053.1 | 6 | 5270 | ||
XM_005245848.3 | XP_005245905.1 | 7 | 5505 | |||
XM_005245849.3 | XP_005245906.1 | 7 | 5610 | |||
XM_005245850.3 | XP_005245907.1 | 8 | 5630 | |||
XM_005245851.3 | XP_005245908.1 | 7 | 5502 | |||
XM_005245852.3 | XP_005245909.1 | 6 | 5279 | |||
XM_011541255.1 | XP_011539557.1 | 7 | 5184 | |||
XM_011541256.2 | XP_011539558.1 | 7 | 5624 | |||
XM_011541257.2 | XP_011539559.1 | 6 | 6379 | |||
GenBank mRNA | AK125431.1 | - | 1 | 1988 | ||
BC002677.2 | - | 1 | 1066 | |||
BC014394.1 | - | 1 | 1066 | |||
U79259.1 | - | 1 | 1031 |