Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPINK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014471.1 | NP_055286.1 | 3, 4 | 215 | ||
GenBank mRNA | AF048700.1 | - | 3, 4 | 215 | ||
BC110068.1 | - | 3, 4 | 215 | 40013921 | ||
EU794680.1 | - | 3, 4 | 215 |