Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CECR6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163079.1 | NP_001156551.1 | 2 | 853 | ||
NM_031890.3 | NP_114096.1 | 1 | 1917 | |||
XM_011546124.2 | XP_011544426.1 | 1 | 1944 | |||
XM_011546125.2 | XP_011544427.1 | 2 | 1263 | |||
GenBank mRNA | AF307451.1 | - | 1 | 1847 | ||
AK095609.1 | - | 2 | 847 | |||
BG472529.1 | - | 1 | 169 |