Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NKIRAS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001349.2 | NP_001001349.1 | 4 | 490 | ||
NM_001144927.1 | NP_001138399.1 | 4 | 594 | |||
NM_001144928.1 | NP_001138400.1 | 4 | 407 | |||
NM_001144929.1 | NP_001138401.1 | 5 | 478 | |||
NM_017595.5 | NP_060065.2 | 4 | 575 | |||
XM_005257251.1 | XP_005257308.1 | 4 | 602 | |||
GenBank mRNA | AF229840.1 | - | 3 | 425 | ||
AK054571.1 | - | 4 | 602 | |||
AK297975.1 | - | 4 | 375 | |||
AL137682.1 | - | 2 | 2315 | |||
BC007316.2 | - | 4 | 527 | |||
BC007450.2 | - | 4 | 483 | 3028708 | ||
BC063498.1 | - | 4 | 1046 | 5248950 | ||
BG717103.1 | - | 4 | 596 |