Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NTMT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286796.1 | NP_001273725.1 | 2 | 369 | ||
NM_001286797.1 | NP_001273726.1 | 2 | 182 | |||
NM_001286798.1 | NP_001273727.1 | 2 | 243 | |||
NM_001286799.1 | NP_001273728.1 | 2 | 172 | |||
NM_001286800.1 | NP_001273729.1 | 2 | 217 | |||
NM_001286801.1 | NP_001273730.1 | 2 | 243 | |||
NM_014064.3 | NP_054783.2 | 2 | 217 | |||
NR_104596.1 | 2 | 217 | ||||
XM_005251939.3 | XP_005251996.1 | 2 | 222 | |||
XM_017014642.1 | XP_016870131.1 | 2 | 139 | |||
GenBank mRNA | AF110776.1 | - | 2 | 157 | ||
AK290957.1 | - | 2 | 214 | |||
AK292332.1 | - | 2 | 219 | |||
AK298840.1 | - | 2 | 207 | |||
AK307056.1 | - | 2 | 208 | |||
AK313829.1 | - | 2 | 207 | |||
BC001396.2 | - | 2 | 196 | 3050940 | ||
BC033234.1 | - | 2 | 171 | 5426447 | ||
BE392289.1 | - | 2 | 174 | |||
BF316784.1 | - | 214 | ||||
BG468453.1 | - | 2 | 379 | |||
BI083998.1 | - | 2 | 182 | |||
BM554138.1 | - | 2 | 172 | |||
BQ923165.1 | - | 2 | 196 | |||
CN406947.1 | - | 2 | 230 |