Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MACROD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014067.3 | NP_054786.2 | 7, 8 | 926 | ||
XM_005273939.4 | XP_005273996.1 | 7, 8 | 1465 | |||
XM_011544970.2 | XP_011543272.1 | 6, 7 | 1435 | |||
GenBank mRNA | AF086206.1 | - | 546 | |||
AF202922.2 | - | 7, 8 | 859 | |||
BC000270.1 | - | 7, 8 | 874 | 3357931 | ||
BC003188.1 | - | 7, 8 | 680 | 3502054 | ||
BC007297.1 | - | 7, 8 | 874 | 3349763 | ||
BC008316.1 | - | 7, 8 | 874 | 3342973 |