Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PYCARD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013258.4 | NP_037390.2 | 3 | 488 | ||
NM_145182.2 | NP_660183.1 | 2 | 431 | |||
GenBank mRNA | AB023416.2 | - | 3 | 488 | ||
AF184073.1 | - | 3 | 476 | |||
AF255794.1 | - | 2 | 419 | |||
AF310103.1 | - | 3 | 402 | |||
AF384665.1 | - | 3 | 455 | |||
AK000211.1 | - | 3 | 440 | |||
AK314422.1 | - | 3 | 480 | |||
BC004470.2 | - | 4 | 458 | 3678925 | ||
BC013569.1 | - | 3 | 395 |