Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLEC2D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004419.4 | NP_001004419.1 | 2 | 116 | ||
NM_001197318.2 | NP_001184247.1 | 2 | 116 | |||
NM_013269.5 | NP_037401.1 | 2 | 116 | |||
NR_036693.2 | 2 | 116 | ||||
GenBank mRNA | AF133299.1 | - | 2 | 102 | ||
AF285087.1 | - | 2 | 94 | |||
AF285088.1 | - | 11 | ||||
AF285089.1 | - | 1 | 26 | |||
AK310150.1 | - | 2 | 112 | |||
AK310554.1 | - | 2 | 107 | |||
AK313554.1 | - | 2 | 107 | |||
BC019883.1 | - | 2 | 96 | 4524891 | ||
BU929933.1 | - | 2 | 101 | |||
CB958277.1 | - | 2 | 113 | |||
DA814760.1 | - | 2 | 111 | |||
DB111977.1 | - | 2 | 116 | |||
DB124897.1 | - | 2 | 112 | |||
FN813349.1 | - | 2 | 89 | |||
KF958454.1 | - | 2 | 89 | |||
KF958455.1 | - | 2 | 89 | |||
KF958456.1 | - | 2 | 89 | |||
KF958457.1 | - | 2 | 89 | |||
KF958458.1 | - | 2 | 89 | |||
KF971856.1 | - | 2 | 89 | |||
KF971857.1 | - | 2 | 89 | |||
KF971859.1 | - | 2 | 89 |