Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UQCR10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003684.1 | NP_001003684.1 | 1 | 158 | ||
NM_013387.3 | NP_037519.2 | 1 | 158 | |||
GenBank mRNA | AB028598.1 | - | 1 | 159 | ||
AF112217.1 | - | 1 | 147 | |||
AF161468.1 | - | 1 | 145 | |||
AF161500.1 | - | 1 | 155 | |||
AF161536.1 | - | 1 | 164 | |||
BC005402.1 | - | 1 | 158 | 3961930 | ||
BC015971.1 | - | 1 | 152 | 4097106 | ||
BI754905.1 | - | 1 | 120 | |||
BU935406.1 | - | 158 | ||||
CA311834.1 | - | 3 | 773 | |||
CB999839.1 | - | 1 | 160 | |||
CD674029.1 | - | 1 | 25 |