Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UQCR10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003684.1 | NP_001003684.1 | 2 | 348 | ||
NM_013387.3 | NP_037519.2 | 2 | 289 | |||
GenBank mRNA | AB028598.1 | - | 2 | 290 | ||
AF112217.1 | - | 2 | 278 | |||
AF161468.1 | - | 2 | 276 | |||
AF161500.1 | - | 2 | 346 | |||
AF161536.1 | - | 2 | 295 | |||
AK225973.1 | - | 2 | 274 | |||
BC005402.1 | - | 2 | 289 | 3961930 | ||
BC015971.1 | - | 2 | 283 | 4097106 | ||
BI754905.1 | - | 2 | 251 | |||
BU935406.1 | - | 348 | ||||
CA311834.1 | - | 2 | 642 | |||
CB999839.1 | - | 2 | 291 | |||
CD674029.1 | - | 2 | 156 |