Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PYCR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271681.1 | NP_001258610.1 | 5 | 728 | ||
NM_013328.3 | NP_037460.2 | 6 | 950 | |||
GenBank mRNA | AF087859.1 | - | 6 | 868 | ||
AF151351.1 | - | 6 | 747 | |||
AK001508.1 | - | 4 | 2329 | |||
AK291913.1 | - | 6 | 785 | |||
BC014868.1 | - | 6 | 841 | 3919161 | ||
BC020553.1 | - | 6 | 875 | 2901360 | ||
BU538765.1 | - | 5 | 603 | |||
KU178615.1 | - | 6 | 720 | |||
KU178616.1 | - | 6 | 544 |