Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NENF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013349.4 | NP_037481.1 | 2 | 253 | ||
GenBank mRNA | AB126219.1 | - | 2 | 196 | ||
AF049672.1 | - | 2 | 51 | |||
AF173937.1 | - | 2 | 256 | |||
AK074431.1 | - | 2 | 221 | |||
AK223135.1 | - | 2 | 221 | |||
AY762102.1 | - | 2 | 212 | |||
BC008823.2 | - | 2 | 202 | 4303614 |