Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NENF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013349.4 | NP_037481.1 | 4 | 526 | ||
NR_026598.1 | 3 | 465 | ||||
GenBank mRNA | AB126219.1 | - | 4 | 469 | ||
AF049672.1 | - | 4 | 324 | |||
AF173937.1 | - | 4 | 529 | |||
AK074431.1 | - | 4 | 494 | |||
AK223135.1 | - | 4 | 494 | |||
AY762102.1 | - | 4 | 485 | |||
BC008823.2 | - | 4 | 475 | 4303614 | ||
DB574171.1 | - | 1 | 36 |