Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OBP2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293189.1 | NP_001280118.1 | 4 | 336 | ||
NM_001293193.1 | NP_001280122.1 | 3 | 202 | |||
NM_014582.2 | NP_055397.1 | 4 | 336 | |||
NR_120603.1 | 4 | 336 | ||||
XM_017014656.1 | XP_016870145.1 | 5 | 1212 | |||
GenBank mRNA | AI219510.1 | - | 4 | 352 | ||
AJ251021.1 | - | 4 | 336 | |||
AJ251022.1 | - | 4 | 336 | |||
AJ251023.1 | - | 4 | 202 | |||
AJ251024.1 | - | 4 | 336 | |||
BC069563.1 | - | 4 | 321 | 7262146 | ||
BC143803.1 | - | 4 | 322 | 9052317 |
Gene Symbol: OBP2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AJ251028.1 | - | 3 | 202 |