Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OBP2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293189.1 | NP_001280118.1 | 5, 6 | 605 | ||
NM_001293193.1 | NP_001280122.1 | 4, 5 | 471 | |||
NM_014582.2 | NP_055397.1 | 5, 6 | 540 | |||
NR_120603.1 | 5, 6 | 589 | ||||
XM_017014656.1 | XP_016870145.1 | 6, 7 | 1481 | |||
GenBank mRNA | AI219510.1 | - | 2, 3 | 148 | ||
AJ251021.1 | - | 5, 6 | 540 | |||
AJ251022.1 | - | 5, 6 | 589 | |||
AJ251023.1 | - | 5, 6 | 471 | |||
AJ251024.1 | - | 5, 6 | 605 | |||
AM087898.1 | - | 5, 6 | 456 | |||
BC069563.1 | - | 5, 6 | 525 | 7262146 |