Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PILRA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013439.2 | NP_038467.2 | 5 | 951 | ||
NM_178272.1 | NP_840056.1 | 4 | 732 | |||
NM_178273.1 | NP_840057.1 | 3 | 698 | |||
GenBank mRNA | AF161080.1 | - | 5 | 951 | ||
AJ400841.1 | - | 5 | 892 | |||
AJ400842.1 | - | 4 | 649 | |||
AJ400843.1 | - | 3 | 615 | |||
BC017812.1 | - | 4 | 719 | 4690881 | ||
BF432841.1 | - | 3 | 369 |