Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLTSCR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015711.3 | NP_056526.3 | 6 | 1055 | ||
XM_005258833.4 | XP_005258890.1 | 6 | 1098 | |||
XM_006723180.3 | XP_006723243.1 | 6 | 1098 | |||
XM_011526882.2 | XP_011525184.1 | 2 | 2065 | |||
XM_011526883.2 | XP_011525185.1 | 6 | 1098 | |||
GenBank mRNA | AF182077.1 | - | 1 | 1071 | ||
BC032065.1 | - | 6 | 1055 |