Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other KCNIP2 SIRNA Assays ›
Gene Symbol:
KCNIP2
Gene Name:
potassium voltage-gated channel interacting protein 2
Gene Aliases:
KCHIP2
Chromosome Location:
Chr.10: 101825971 - 101843920 on Build GRCh38
UniGene ID:
Hs.97044
Species:
Human
siRNA ID
HSS147163
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: KCNIP2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_014591.4
NP_055406.2
8
1010
NM_173191.2
NP_775283.1
8
965
NM_173192.2
NP_775284.1
7
911
NM_173193.2
NP_775285.1
6
836
NM_173194.2
NP_775286.1
6
492
NM_173195.2
NP_775287.1
6
815
NM_173197.2
NP_775289.1
5
707
XM_005269729.2
XP_005269786.1
7
813
XM_005269730.2
XP_005269787.1
6
770
XM_006717812.2
XP_006717875.1
8
867
XM_011539731.2
XP_011538033.1
6
507
XM_017016161.1
XP_016871650.1
6
562
GenBank mRNA
AF199598.1
-
7
559
AF295076.1
-
6
463
AF295530.1
-
6
484
AF347114.1
-
6
463
AF367018.1
-
8
658
AF367019.1
-
5
355
AF367020.1
-
4
334
AF367021.1
-
8
613
AK027347.1
-
6
483
AK055089.1
-
8
889
AK094668.1
-
10
1017
AK125382.1
-
6
1522
AK295324.1
-
4
446
AK301433.1
-
6
484
AK309368.1
-
6
691
AK314536.1
-
6
696
AK315042.1
-
7
791
AL136722.1
-
6
510
AY026328.1
-
6
463
AY302141.1
-
6
451
BC034685.1
-
7
774
4155871
DQ148480.1
-
8
613
DQ148481.1
-
7
559
DQ148482.1
-
6
463
DQ148483.1
-
6
478
DQ148484.1
-
5
405
DQ862463.1
-
6
463
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
detection of calcium ion
potassium ion transport
muscle contraction
signal transduction
chemical synaptic transmission
regulation of heart contraction
clustering of voltage-gated potassium channels
cardiac conduction
potassium ion export
potassium ion transmembrane transport
membrane repolarization
regulation of potassium ion transmembrane transport
regulation of cation channel activity
A-type (transient outward) potassium channel activity
calcium ion binding
protein binding
potassium channel regulator activity
ion channel binding
ER retention sequence binding
protein N-terminus binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-8skhh:80/10.239.164.194:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline