Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EXOSC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002269.2 | NP_001002269.1 | 1 | 111 | ||
NM_016042.3 | NP_057126.2 | 1 | 111 | |||
GenBank mRNA | AF151860.1 | - | 1 | 36 | ||
AF281132.1 | - | 1 | 87 | |||
AK289571.1 | - | 1 | 80 | |||
AK290864.1 | - | 1 | 76 | |||
BC002437.2 | - | 1 | 75 | 3346075 | ||
BC008880.2 | - | 1 | 65 | 4300826 | ||
BP237222.1 | - | 1 | 110 | |||
CF994051.1 | - | 1 | 119 | |||
DA352003.1 | - | 1 | 85 |