Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAM16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016069.9 | NP_057153.8 | 2 | 165 | ||
GenBank mRNA | AF151894.1 | - | 2 | 72 | ||
AF349455.1 | - | 2 | 59 | |||
AK026514.1 | - | 2 | 92 | |||
BC005024.1 | - | 2 | 74 | 3955475 | ||
BG715429.1 | - | 2 | 169 | |||
CR457362.1 | - | 1 | 27 |
Gene Symbol: CORO7-PAM16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201479.1 | NP_001188408.1 | 28 | 3239 | ||
GenBank mRNA | AL833954.1 | - | 30 | 3314 |