Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C5orf45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017987.2 | NP_001017987.1 | 1, 2 | 101 | ||
NM_016175.3 | NP_057259.2 | 1, 2 | 101 | |||
GenBank mRNA | AK293901.1 | - | 1, 2 | 67 | ||
AK295104.1 | - | 1, 2 | 94 | |||
AK298954.1 | - | 1, 2 | 58 | |||
BC056889.1 | - | 1, 2 | 54 | |||
BC069051.1 | - | 1, 2 | 69 | 3945859 | ||
BF965313.1 | - | 1, 2 | 102 | |||
BQ642639.1 | - | 1, 2 | 57 | |||
BX537968.1 | - | 1, 2 | 122 | |||
BX647375.1 | - | 1, 2 | 69 | |||
CN363239.1 | - | 1, 2 | 25 | |||
CR936785.1 | - | 1, 2 | 122 |