Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016176.3 | NP_057260.2 | 4 | 884 | ||
NM_016547.2 | NP_057631.1 | 4 | 884 | |||
XM_011541556.1 | XP_011539858.1 | 4 | 822 | |||
GenBank mRNA | AF132749.1 | - | 4 | 798 | ||
AF153686.1 | - | 4 | 848 | |||
AF178986.1 | - | 4 | 848 | |||
AK000717.1 | - | 2 | 226 | |||
AK027277.1 | - | 5 | 476 | |||
AK075352.1 | - | 4 | 818 | |||
AK222521.1 | - | 4 | 802 | |||
AK223079.1 | - | 4 | 800 | |||
AK299810.1 | - | 4 | 758 | |||
BC006211.2 | - | 4 | 765 | 3162764 | ||
BC007625.1 | - | 3 | 633 | 3503540 | ||
BC008917.2 | - | 4 | 784 | 3622725 | ||
BC011244.1 | - | 4 | 738 | 4153655 | ||
BC022375.1 | - | 4 | 818 | 4839718 | ||
L79912.1 | - | 3 | 563 |