Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002032.2 | NP_001002032.1 | 3 | 568 | ||
NM_001002033.2 | NP_001002033.1 | 4 | 663 | |||
NM_001288609.1 | NP_001275538.1 | 3 | 1252 | |||
NM_001288610.1 | NP_001275539.1 | 3 | 615 | |||
NM_001288611.1 | NP_001275540.1 | 3 | 568 | |||
NM_016185.3 | NP_057269.1 | 3 | 568 | |||
NR_109933.1 | 3 | 568 | ||||
GenBank mRNA | AF086910.2 | - | 3 | 568 | ||
AF177862.1 | - | 3 | 385 | |||
AF266846.1 | - | 3 | 397 | |||
AK223321.1 | - | 3 | 397 | |||
AK308253.1 | - | 3 | 615 | |||
AK312613.1 | - | 3 | 397 | |||
AY322169.1 | - | 3 | 560 | |||
AY322170.1 | - | 4 | 655 | |||
BC001420.2 | - | 3 | 386 | 3140086 | ||
BC039343.1 | - | 3 | 1254 | 5269120 | ||
BG779109.1 | - | 3 | 397 | |||
DA604735.1 | - | 3 | 397 |