Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VPS28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016208.3 | NP_057292.1 | 9 | 594 | ||
NM_183057.2 | NP_898880.1 | 9 | 594 | |||
XM_005272323.3 | XP_005272380.1 | 9 | 535 | |||
XM_005272324.3 | XP_005272381.1 | 9 | 535 | |||
XM_005272325.3 | XP_005272382.1 | 9 | 730 | |||
XM_011517099.1 | XP_011515401.1 | 9 | 648 | |||
XM_011517100.1 | XP_011515402.1 | 9 | 648 | |||
XM_017013529.1 | XP_016869018.1 | 9 | 730 | |||
GenBank mRNA | AF182844.1 | - | 8 | 546 | ||
AF316887.1 | - | 8 | 528 | |||
AF445417.1 | - | 1 | 1139 | |||
AK026865.1 | - | 9 | 544 | |||
AK090942.1 | - | 7 | 1775 | |||
AK091979.1 | - | 5 | 1992 | |||
AK129716.1 | - | 1364 | ||||
BC006485.1 | - | 8 | 530 | 3535787 | ||
BC019321.1 | - | 8 | 530 | 2822758 | ||
BC050713.2 | - | 9 | 542 | 5741129 | ||
KU178635.1 | - | 8 | 485 | |||
KU178636.1 | - | 8 | 434 |