Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIGP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320480.1 | NP_001307409.1 | 5 | 703 | ||
NM_016430.3 | NP_057514.2 | 6 | 717 | |||
NM_153681.2 | NP_710148.1 | 4 | 658 | |||
NM_153682.2 | NP_710149.1 | 5 | 551 | |||
XM_005260990.4 | XP_005261047.1 | 5 | 416 | |||
XM_011529596.2 | XP_011527898.1 | 5 | 450 | |||
XM_017028364.1 | XP_016883853.1 | 5 | 468 | |||
XM_017028365.1 | XP_016883854.1 | 6 | 659 | |||
GenBank mRNA | AB035742.1 | - | 5 | 483 | ||
AB035743.1 | - | 5 | 413 | |||
AB035744.1 | - | 5 | 445 | |||
AB035745.1 | - | 4 | 465 | |||
AB037162.1 | - | 5 | 487 | |||
AB037163.1 | - | 4 | 456 | |||
AB037164.1 | - | 5 | 568 | |||
AB039659.1 | - | 4 | 370 | |||
AB451328.1 | - | 3 | 292 | |||
AB451472.1 | - | 3 | 292 | |||
AF216305.1 | - | 6 | 633 | |||
AF237812.1 | - | 4 | 507 | |||
AI127387.1 | - | 1 | 250 | |||
AK026311.1 | - | 4 | 443 | |||
AK314457.1 | - | 4 | 451 | |||
AK316609.1 | - | 4 | 388 | |||
BC005180.1 | - | 5 | 432 | 3929930 | ||
BC011007.1 | - | 5 | 436 | 4281552 | ||
BT007053.1 | - | 4 | 370 |