Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COX16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204090.1 | NP_001191019.1 | 1 | 184 | ||
NM_016468.6 | NP_057552.1 | 1 | 184 | |||
GenBank mRNA | AF151037.1 | - | 1 | 131 | ||
AF226729.1 | - | 1 | 158 | |||
AK290743.1 | - | 1 | 132 | |||
BC001702.2 | - | 1 | 84 | 2966935 | ||
BM799209.1 | - | 1 | 153 | |||
CB156527.1 | - | 1 | 128 | |||
CR457199.1 | - | 1 | 40 | |||
FJ460510.1 | - | 1 | 184 | |||
FJ460511.1 | - | 1 | 184 |