Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC174 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016474.4 | NP_057558.3 | 5 | 418 | ||
NR_135523.1 | 5 | 418 | ||||
XM_017006554.1 | XP_016862043.1 | 5 | 481 | |||
XM_017006555.1 | XP_016862044.1 | 5 | 479 | |||
GenBank mRNA | AF151046.1 | - | 5 | 381 | ||
AK309525.1 | - | 5 | 391 | |||
BC013999.2 | - | 5 | 394 | 3936914 | ||
BC033897.1 | - | 5 | 405 | 4812977 | ||
BP195134.1 | - | 5 | 418 | |||
CD580179.2 | - | 5 | 416 |