Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC174 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016474.4 | NP_057558.3 | 7 | 708 | ||
NR_135523.1 | 7 | 708 | ||||
XM_017006554.1 | XP_016862043.1 | 7 | 771 | |||
XM_017006555.1 | XP_016862044.1 | 7 | 769 | |||
GenBank mRNA | AK309525.1 | - | 7 | 681 | ||
BC013999.2 | - | 7 | 684 | 3936914 | ||
BC033897.1 | - | 7 | 695 | 4812977 |