Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC174 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016474.4 | NP_057558.3 | 8 | 827 | ||
GenBank mRNA | BC013999.2 | - | 8 | 803 | 3936914 | |
BC033897.1 | - | 8 | 814 | 4812977 |