Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM178B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122646.2 | NP_001116118.2 | 16 | 2225 | ||
NM_001172667.1 | NP_001166138.1 | 4 | 426 | |||
NM_016490.4 | NP_057574.2 | 4 | 544 | |||
XM_011511294.2 | XP_011509596.1 | 15 | 2153 | |||
XM_017004263.1 | XP_016859752.1 | 17 | 2390 | |||
XM_017004264.1 | XP_016859753.1 | 17 | 2351 | |||
XM_017004266.1 | XP_016859755.1 | 7 | 798 | |||
XM_017004267.1 | XP_016859756.1 | 4 | 403 | |||
XM_017004268.1 | XP_016859757.1 | 4 | 429 | |||
GenBank mRNA | AK122705.1 | - | 10 | 1104 | ||
AK308698.1 | - | 10 | 1107 | |||
BC038583.1 | - | 4 | 505 | |||
BC039488.1 | - | 14 | 1492 | |||
BC051379.1 | - | 4 | 544 | |||
BX103404.1 | - | 4 | 404 |