Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ANKMY1 SIRNA Assays ›
Gene Symbol:
ANKMY1
Gene Name:
ankyrin repeat and MYND domain containing 1
Gene Aliases:
ZMYND13
Chromosome Location:
Chr.2: 240468632 - 240561110 on Build GRCh38
UniGene ID:
Hs.656615
Species:
Human
siRNA ID
HSS147381
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ANKMY1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001282771.1
NP_001269700.1
14
2720
NM_001282780.1
NP_001269709.1
10
1790
NM_001282781.1
NP_001269710.1
13
1736
NM_001308375.1
NP_001295304.1
11
2161
NM_016552.3
NP_057636.2
13
2557
NM_017844.3
NP_060314.2
10
1781
XM_011511300.2
XP_011509602.1
14
2904
XM_011511301.1
XP_011509603.1
14
2904
XM_011511304.2
XP_011509606.1
13
2763
XM_011511305.2
XP_011509607.1
14
2904
XM_011511309.2
XP_011509611.1
14
2904
XM_011511313.2
XP_011509615.1
14
2933
XM_011511317.1
XP_011509619.1
10
1974
XM_011511318.1
XP_011509620.1
10
1965
XM_011511322.1
XP_011509624.1
12
2214
XM_017004271.1
XP_016859760.1
14
2904
XM_017004272.1
XP_016859761.1
13
2796
XM_017004273.1
XP_016859762.1
13
2772
XM_017004274.1
XP_016859763.1
13
2763
XM_017004275.1
XP_016859764.1
14
2670
XM_017004276.1
XP_016859765.1
15
2991
XM_017004277.1
XP_016859766.1
12
2664
XM_017004278.1
XP_016859767.1
12
2655
XM_017004279.1
XP_016859768.1
11
2523
XM_017004280.1
XP_016859769.1
13
2763
XM_017004281.1
XP_016859770.1
13
2510
XM_017004282.1
XP_016859771.1
13
2754
XM_017004283.1
XP_016859772.1
15
2991
XM_017004284.1
XP_016859773.1
12
2214
XM_017004285.1
XP_016859774.1
12
1980
XM_017004286.1
XP_016859775.1
12
2613
XM_017004287.1
XP_016859776.1
10
2373
XM_017004288.1
XP_016859777.1
11
2064
XM_017004289.1
XP_016859778.1
12
2613
XM_017004290.1
XP_016859779.1
12
2281
XM_017004291.1
XP_016859780.1
13
2583
GenBank mRNA
AB034636.1
-
13
2487
AK000506.1
-
10
1748
AK225534.1
-
10
1748
AK314534.1
-
14
2704
AL833866.1
-
10
1637
BC073146.1
-
10
1731
5743806
CR936759.1
-
11
1830
Back To Top
More Information
Gene Ontology Categories:
Process(es)
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-x5whk:80/10.239.163.84:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0