Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THEG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016585.4 | NP_057669.1 | 6 | 733 | ||
NM_199202.2 | NP_954672.1 | 5 | 661 | |||
XM_011528049.2 | XP_011526351.1 | 6 | 1412 | |||
XM_011528050.2 | XP_011526352.1 | 5 | 1339 | |||
XM_011528051.2 | XP_011526353.1 | 3 | 1209 | |||
GenBank mRNA | AB033129.1 | - | 6 | 733 | ||
AF268610.1 | - | 6 | 699 | |||
BC028574.2 | - | 6 | 712 | 4821678 | ||
DR731298.1 | - | 2 | 118 |